missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ COL9A1 Recombinant Protein

Brand:  Abnova™ H00001297-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16153391

  • £425.00 / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Antibody Production, ELISA, Protein Array, Western Blot
61.82
8
Glutathione Sepharose 4 Fast Flow
25μg
-80°C
Recombinant
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human COL9A1 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers