missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents

Brand:  Fisher Bioreagents BP2510-25

Code : 32

Additional Details : CAS Number : 18439-24-2 Weight : 0.05003kg

Packaging Amber Glass

 View more versions of this product

Product Code. 11473064

  • £80.30 / 25mg


Estimated Shipment
20-03-2018
Add to basket

Specifications

Chemical Identifiers

Coenzyme A
18439-24-2
≥96 %

Specifications

Coenzyme A
Pass Test
≥96 %
96% min.
White
18439-24-2
Powder/Solid
0.80 to 0.89
Amber Glass
25mg
Videos
SDS Documentation

Technical Documents