missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COCO Rat anti-Mouse, Clone: 340531, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | COCO/DAND5 |
---|---|
Clone | 340531 |
Immunogen | E. coli-derived recombinant mouse COCO Arg24-Leu185 Accession # Q76LW6 |
Gene ID (Entrez) | 199699 |
Gene Accession No. | Q76LW6 |
Description
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Mouse samples. It is validated for Western Blot.Specifications
COCO/DAND5 | |
E. coli-derived recombinant mouse COCO Arg24-Leu185 Accession # Q76LW6 | |
Q76LW6 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
340531 | |
199699 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) COCO, rhDAN, recombinant mouse (rm) Gremlin, rmCerberus, and recombinant chicken Caronte is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only