missing translation for 'onlineSavingsMsg'
Learn More

Cobaltocene, 98%, ACROS Organics™

Brand:  Acros Organics 405320100

Code : 29

Additional Details : CAS Number : 1277-43-6 Weight : 0.06000kg Transport : UN number : 1325 Chem class : 1 Pack group : II


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10350184

  • £211.50 / 10g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

Cobaltocene
C10H10Co
MFCD00013749
98%
189.12
Bis(cyclopentadienyl)cobalt

Specifications

Cobaltocene
97.5% min.
Metallic Crystals
C10H10Co
MFCD00013749
Bis(cyclopentadienyl)cobalt
10g
1277-43-6
Authentic
98%
189.12
Glass bottle
Black
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cobaltocene

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS