missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni <0.15%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Cobalt titanium oxide shows magnetic properties at room temperature and used in electronics and as spintronic devices. It is used to improve the gas sensing ability of hydrogen sulfide by hydrothermal method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water. |
Formula Weight | 154.83 |
Percent Purity | 99.8% |
Assay | (metals basis excluding Ni), Ni <0.15% |
Chemical Name or Material | Cobalt titanium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'