missing translation for 'onlineSavingsMsg'
Learn More

Cobalt tetramethoxyphenylporphyrin, 94%, ACROS Organics™

Brand:  Acros Organics 405340250

Code : 29

Additional Details : CAS Number : 28903-71-1 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10703624

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Cobalt tetramethoxyphenylporphyrin
≥92%
C48H36CoN4O4
MFCD00010724
28903-71-1
94%
791.77

Specifications

Cobalt tetramethoxyphenylporphyrin
Authentic
≥92%
C48H36CoN4O4
MFCD00010724
Purple to violet
28903-71-1
Crystals or Powder
94%
791.77
Glass bottle
25g
Videos
SDS