Learn More
Cobalt tetramethoxyphenylporphyrin, 94%, ACROS Organics™
Brand: Acros Organics 405340050
Code : 29
Additional Details : CAS Number : 28903-71-1 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
Cobalt tetramethoxyphenylporphyrin | |
94% | |
C48H36CoN4O4 | |
MFCD00010724 |
28903-71-1 | |
94% | |
791.77 |
Specifications
Cobalt tetramethoxyphenylporphyrin | |
Authentic | |
94% | |
C48H36CoN4O4 | |
MFCD00010724 | |
Purple to violet |
28903-71-1 | |
Crystals or Powder | |
94% | |
791.77 | |
Glass bottle | |
5g |