missing translation for 'onlineSavingsMsg'
Learn More

Cobalt powder, -22 mesh, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11310779
Click to view available options
Quantity:
5 g
25 g
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.99% (metals basis)
Quantity 5 g
Mesh Size -22 Mesh
Chemical Name or Material Cobalt powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'