Learn More
Alfa Aesar™ Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina
Brand: Alfa Aesar™ 045579.14
Code : NEW
Additional Details : Weight : 0.02500kg Transport : UN number : 3077 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
Cobalt oxide, Molybdenum oxide | |
On alumina | |
MFCD01074445 | |
UN3077 |
Specifications
Cobalt oxide, Molybdenum oxide | |
Trilobe Extrudate | |
UN3077 | |
Odorless | |
Blue |
On alumina | |
MFCD01074445 | |
Co oxide:typically 3.4 to 4.5%; Mo oxide: typically 11.5 to 14.5% | |
2.5mm | |
25g |