missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g, Thermo Scientific Chemicals

Catalog Number 15487405
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Co 1000µg/g
CAS 7697-37-2
Quantity 50 g
Molecular Formula HNO3
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Cobalt, Oil based standard solution
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'