missing translation for 'onlineSavingsMsg'
Learn More

Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 15457185
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 5 g
Percent Purity 99.8%
Odor Odorless
Particle Size APS 25 to 30nm, Co3O4 layer ≈2nm
Form Powder
Assay (metals basis)
Chemical Name or Material Cobalt nanopowder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'