missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis)

Brand:  Alfa Aesar™ 010694.03

UNSPSC : 12141710

Code : NEW

Additional Details : CAS Number : 10026-22-9 Weight : 0.00100kg Transport : UN number : 1477 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11370949

  • £35.09 / 1g


Estimated Shipment date
from Supplier 11-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Cobalt(II) nitrate hexahydrate
Puratronic™
MFCD00149647
14,2444
10026-22-9
291.04 (182.94 Anhydrous)
UN1477

Specifications

Cobalt(II) nitrate hexahydrate
Puratronic™
99.999%
291.04 (182.94 Anhydrous)
UN1477
1.87g/mL
1.52
55° to 56°C
10026-22-9
Wet Crystalline Aggregates
(metals basis)
MFCD00149647
14,2444
Odorless
Red
1g
Documentation

Technical Documents