missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number 11307888
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10026-22-9
CoH12N2O12
291.03
MFCD00149647
QGUAJWGNOXCYJF-UHFFFAOYSA-N
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
24821
CHEBI:86214
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10026-22-9
CoH12N2O12
291.03
MFCD00149647
QGUAJWGNOXCYJF-UHFFFAOYSA-N
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
24821
CHEBI:86214
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

For Use As a Battery Additive, in the Manufacture of Cobalt Pigments and Invisible Inks, For Decorating Stoneware and Porcelain

Used as a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and porcelain, and in the preparation of catalysts. It is commonly reduced to metallic cobalt or precipitated on various substrates for gischer-tropsch catalysis. It is used as analytical reagent to determine potassium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:
  • As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts
TRUSTED_SUSTAINABILITY
Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0-102.0%
Solubility Information Very soluble in water,alcohol,and most organic solvents.
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
Formula Weight 291.04 (182.94 Anhydrous)
Grade ACS
Odor Odorless
Refractive Index 1.52
Appearance Red crystalline powder
Density 1.87 g/cm3
Chemical Name or Material Cobalt(II) nitrate hexahydrate, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'