missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
Used as a battery additive, in the manufacture of cobalt pigments and invisible inks, for decorating stoneware and porcelain, and in the preparation of catalysts. It is commonly reduced to metallic cobalt or precipitated on various substrates for gischer-tropsch catalysis. It is used as analytical reagent to determine potassium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications:
- As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts
Specifications
Melting Point | 55°C to 56°C |
Color | Red |
Physical Form | Crystalline |
Quantity | 2.5 kg |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Very soluble in water,alcohol,and most organic solvents. |
IUPAC Name | λ²-cobalt(2+) hexahydrate dinitrate |
Formula Weight | 291.04 (182.94 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'