missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cobalt(II) molybdenum oxide, 99.9% (metals basis), Ni 0.5% max

Brand:  Alfa Aesar™ 041376.09

Code : NEW

Additional Details : CAS Number : 13762-14-6 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11334088

  • £26.71 / 10g


Estimated Shipment date
from Supplier 19-06-2018
Add to basket

Specifications

Chemical Identifiers

Cobalt(II) molybdenum oxide
218.87
UN3077
13762-14-6
MFCD00016019
Cobalt(II) molybdate

Specifications

Cobalt(II) molybdenum oxide
Powder
(metals basis)
MFCD00016019
Cobalt(II) molybdate
−325 Mesh
10g
13762-14-6
99.9%
218.87
UN3077
Ni 0.5% max.
Green
Videos
SDS