Learn More
Thermo Scientific Alfa Aesar Cobalt(II) molybdenum oxide, 99.9% (metals basis), Ni 0.5% max
Brand: Thermo Scientific Alfa Aesar 041376.18
Cobalt(II) molybdenum oxide, 99.9% (metals basis), Ni 0.5% max
Code : NEW
Additional Details : CAS Number : 13762-14-6 Weight : 0.05000kg Transport : UN number : 3077 Chem class : 9 Pack group : III