missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) isopropoxide, Thermo Scientific Chemicals
46.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Cobalt(II) isopropoxide is used in display & imaging, catalysis & synthesis and ferroelectric metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Packaging | Ampule |
Quantity | 1 g |
UN Number | UN3181 |
Sensitivity | Moisture sensitive |
Solubility Information | It reacts with water. |
Formula Weight | 178.11 |
Physical Form | Powder |
Chemical Name or Material | Cobalt (II) isopropoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'