missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Chloride Hexahydrate, Reagent grade, Honeywell™

Catalog Number p-7120333
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7791-13-1
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
This item is not returnable. View return policy
7791-13-1
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Melting Point 86°C
Color Red
Quantity 25 g
Linear Formula CoCl2 · 6H2O
UN Number UN3077
Formula Weight 237.93g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 40mmHg (0°C)
Density 1.9g/cu.cm
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'