missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals

Catalog Number 10454321
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
7791-13-1
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
This item is not returnable. View return policy
7791-13-1
Cl2CoH12O6
237.92
MFCD00149652
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 86.0°C
Color Lilac
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15, 2421
Solubility Information Solubility in water: 970g/L (20°C)
Formula Weight 237.93
Percent Purity 98 to 102%
Grade ACS Reagent
Packaging Glass bottle
Vapor Pressure Negligible
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'