missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11313888
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7789-43-7
Br2Co
218.741
MFCD00016017
BZRRQSJJPUGBAA-UHFFFAOYSA-L
cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
24610
dibromocobalt
[Co](Br)Br
This item is not returnable. View return policy
7789-43-7
Br2Co
218.741
MFCD00016017
BZRRQSJJPUGBAA-UHFFFAOYSA-L
cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
24610
dibromocobalt
[Co](Br)Br

Used primarily as a catalyst in some processes. Chiefly used in hygrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 678°C (in N2)
Color Green
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Soluble in methyl acetate,ether,alcohol,acetone.
Formula Weight 218.75
Sensitivity Hygroscopic
Appearance Green (powder/flake/crystalline/beads, etc.)
Density 4.909 g/cm3
Chemical Name or Material Cobalt(II) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'