missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) acetylacetonate, 99%, Thermo Scientific Chemicals
37.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
29.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
29.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165.0°C to 170.0°C |
Color | Purple |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5 to 101.5% (ex Co) |
Packaging | Glass bottle |
Linear Formula | (CH3COCH=C(CH3)O)2Co |
Quantity | 50 g |
Beilstein | 01, 783 |
Fieser | 17,87 |
Solubility Information | Solubility in water: 5g/L (20°C). Other solubilities: 7g/kg alcohol (20°C),173g/kg benzene (20°C) |
Show More |
Safety and Handling
Product Identifier
- Cobalt(II) acetylacetonate
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304-IF INHALED:.
- P340-Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'