missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, 99% (m.b.), packaged under Ar in resealable ChemSeal bottles, Thermo Scientific Chemicals

Catalog Number p-7058588
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
142600-61-1
C6H14CoO4
209.107
MFCD00798554
BFBNUELICQBJBC-UHFFFAOYSA-N
cobalt ii 2-methoxyethoxide
131873636
cobalt(2+);2-methoxyethanolate
COCC[O-].COCC[O-].[Co+2]
This item is not returnable. View return policy
142600-61-1
C6H14CoO4
209.107
MFCD00798554
BFBNUELICQBJBC-UHFFFAOYSA-N
cobalt ii 2-methoxyethoxide
131873636
cobalt(2+);2-methoxyethanolate
COCC[O-].COCC[O-].[Co+2]

Used as a nano-materials, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Packaging ChemSeal™ bottles
Linear Formula Co(OCH2CH2OCH3)2
Quantity 25 mL
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information Denatured by water. Miscible with alcohols,ketones and esters.
Formula Weight 209.11
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Cobalt (II) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'