missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.127mm (0.005in) thick, 50x50mm (2x2in), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 15496305
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
Percent Purity 99.9%
Odor Odorless
Weight ≈2.83g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Cobalt foil, 0.127mm (0.005 in.) thick, 50x50mm (2x2 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'