missing translation for 'onlineSavingsMsg'
Learn More

Coal tar, Thermo Scientific Chemicals

Catalog Number p-7052075
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8007-45-2
MFCD00146482
Tar
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8007-45-2
MFCD00146482
Tar

Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Petroleum-like
Quantity 100 mL
Merck Index 14,2426
Solubility Information Not miscible or difficult to mix with water.
Physical Form Viscous Liquid
Chemical Name or Material Coal tar

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'