missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Coagulation Factor III/Tissue Factor Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Coagulation Factor III/Tissue Factor |
---|---|
Clone | 355220 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot.Specifications
Coagulation Factor III/Tissue Factor | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P20352 | |
2152 | |
Mouse myeloma cell line NS0-derived recombinant mouse Coagulation Factor III Ala29-Glu251 Accession # P20352 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. |
355220 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
IgG2c | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only