Learn More
Co 102862, Tocris Bioscience™
State-dependent of voltage-gated sodium channels
Brand:  Tocris (Bio-Techne) 264210
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13441634
  • £116.92 / 10mg
Estimated Shipment
14-09-2020
Description
Description
Broad spectrum, state-dependent blocker of voltage-gated sodium channels. Displays ∽ 80-fold higher affinity for inactivated Na+ channels compared to channels in the resting state. Anticonvulsant; displays activity in rodent models of tonic/clonic and partial-complex seizures.
Specifications
Chemical Identifiers
181144-66-1
273.267
2-4-4-fluorophenoxy benzylidene hydrazinecarboxamide, hydrazinecarboxamide, 2-4-4-fluorophenoxy phenyl methylene, e-4-4-fluorophenoxy phenyl methyleneamino urea, 4-4-fluorophenoxy phenyl methylidene aminourea
[[4-(4-fluorophenoxy)phenyl]methylideneamino]urea
C14H12FN3O2
MHUUDVZSPFRUSK-UHFFFAOYSA-N
44483319
C1=CC(=CC=C1C=NNC(=O)N)OC2=CC=C(C=C2)F
Specifications
Co 102862
181144-66-1
2-4-4-fluorophenoxy benzylidene hydrazinecarboxamide, hydrazinecarboxamide, 2-4-4-fluorophenoxy phenyl methylene, e-4-4-fluorophenoxy phenyl methyleneamino urea, 4-4-fluorophenoxy phenyl methylidene aminourea
C1=CC(=CC=C1C=NNC(=O)N)OC2=CC=C(C=C2)F
273.267
273.26
10mg
C14H12FN3O2
MHUUDVZSPFRUSK-UHFFFAOYSA-N
[[4-(4-fluorophenoxy)phenyl]methylideneamino]urea
44483319
>99%