missing translation for 'onlineSavingsMsg'
Learn More

CNQX disodium salt, Tocris Bioscience™

Potent non-NMDA iGluR antagonist; more water soluble form of CNQX (Cat. No. 0190)

Brand:  Tocris (Bio-Techne) 1045/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13468373

  • £508.62 / 50mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

More water soluble disodium salt of the AMPA/kainate antagonist CNQX (Cat. No. 0190). Also available as part of the Kainate Receptor Tocriset™ .
Specifications

Chemical Identifiers

479347-85-8
276.119
cnqx disodium salt, disodium 6-cyano-7-nitroquinoxaline-2,3-diolate, 6-cyano-7-nitroquinoxaline-2,3-dione disodium, sodium 6-cyano-7-nitroquinoxaline-2,3-bis olate, cnqx disodium salt hydrate, 6-quinoxalinecarbonitrile,1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 6-cyano-7-nitroquinoxaline-2,3-dione disodium salt hydrate, 6-cyano-7-nitroquinoxaline-2,3-dione 2na, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxoquinoxaline-6-carbonitrile disodium
disodium;6-cyano-7-nitroquinoxaline-2,3-diolate
C9H2N4Na2O4
YCXDDPGRZKUGDG-UHFFFAOYSA-L
6093155
C1=C(C(=CC2=C1N=C(C(=N2)[O-])[O-])[N+](=O)[O-])C#N.[Na+].[Na+]

Specifications

50mg
479347-85-8
cnqx disodium salt, disodium 6-cyano-7-nitroquinoxaline-2,3-diolate, 6-cyano-7-nitroquinoxaline-2,3-dione disodium, sodium 6-cyano-7-nitroquinoxaline-2,3-bis olate, cnqx disodium salt hydrate, 6-quinoxalinecarbonitrile,1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 6-cyano-7-nitroquinoxaline-2,3-dione disodium salt hydrate, 6-cyano-7-nitroquinoxaline-2,3-dione 2na, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxoquinoxaline-6-carbonitrile disodium
C1=C(C(=CC2=C1N=C(C(=N2)[O-])[O-])[N+](=O)[O-])C#N.[Na+].[Na+]
276.119
276.12
CNQX disodium salt
C9H2N4Na2O4
YCXDDPGRZKUGDG-UHFFFAOYSA-L
disodium;6-cyano-7-nitroquinoxaline-2,3-diolate
6093155
>99%
Videos
Documents