missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CNP Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | CNP/NPPC |
---|---|
Clone | 462608 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
CNP/NPPC Monoclonal specifically detects CNP/NPPC in Human samples. It is validated for Western Blot.Specifications
CNP/NPPC | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P23582 | |
4880 | |
E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human pro-CNP and mature CNP53 and CNP29 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ANP or rhBNP is observed. |
462608 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC | |
NPPC | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only