Learn More
Tocris Bioscience™ CMPD-1
Selective inhibitor of p38α-mediated MK2a phosphorylation; also tubulin polymerization inhibitor
Brand:  Tocris Bioscience™ 218610
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13450744
  • £183.75 / 10mg
Estimated Shipment date
from Supplier 24-02-2020
Description
Description
Non-ATP-competitive, selective inhibitor of p38α-mediated MK2a (mitogen-activated protein kinase-2a) phosphorylation (apparent Ki = 330 nM). Does not inhibit p38α-mediated phosphorylation of MBP and ATF-2. Also tubulin polymerization inhibitor. Cytotoxic in glioblastoma cells at concentrations that do not affect the MK2 pathway.
Specifications
Specifications
CMPD-1
>99
10mg
CMPD-1
349.4