Learn More
CMKLR1 Rat anti-Human, Clone: BZ332, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-7581-80
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218087
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The BZ332 antibody reacts with human chemokine-like receptor 1, CMKLR1. CMKLR1 (also known as ChemR23 or DEZ), is a recently-described serpentine chemoattractant receptor. CMKLR1 has been shown to be expressed by dendritic cells (DC) generated in vitro from monocytes, and to mediate their migration to the proteolytically regulated chemoattractant chemerin. CMKLR1 is expressed by circulating DCs in human blood. Importantly, it is selectively expressed by immature plasmacytoid DCs, but not myeloid DCs, thus enabling discrimination between these two dendritic cell lineages. While CMKLR1 is expressed by circulating pDCs in human blood, mDCs, as well as lymphocytes, monocytes, neutrophils, and eosinophils are negative. Human serum contains a chemoattractant for CMKLR1 which is identical with chemerin; this activity is triggered during blood coagulation, suggesting a mechanism for attraction of pDCs to sites of tissue damage and bleeding. Expression of CMKLR1 and migration to locally activated chemerin may allow the rapid and differential recruitment of pDCs in vivo. Unlike mDCs, circulating pDCs do not respond to inflammatory chemokines.
Specifications
Specifications
CMKLR1
BZ332
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
serpentine chemokine-like receptor 1
IgG2a