missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Clusterin-like 1/CLUL1 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Clusterin-like 1/CLUL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Clusterin-like 1/CLUL1 Polyclonal specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blot.Specifications
Clusterin-like 1/CLUL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CLUL1, Clusterin like 1, clusterin-like 1 (retinal), clusterin-like protein 1, RA337M, Retinal-specific clusterin-like protein | |
CLUL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q15846 | |
27098 | |
Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1 Cys20-Ala442 Accession # Q15846 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Clusterin-like 1/CLUL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Clusterin is observed. |
For Research Use Only