missing translation for 'onlineSavingsMsg'
Learn More

Clostridium Difficile Toxin A Antibody (PCG4.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 18 publications

Brand:  Novus Biologicals NB600-1066-0.1mg

226.10 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18768593

  • £282.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Clostridium Difficile Toxin A Monoclonal specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clostridium Difficile Toxin A
Monoclonal
1.16 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
tcdA, toxA
Full length protein A (C. difficile).
0.1 mg
Immunology
C. difficile Toxin A. Does not cross react with C. difficile Toxin B
Bacteria, Hamster
Purified
Western Blot, ELISA
PCG4.1
Unconjugated
10mM Sodium Phosphate (pH 7.4) and 0.15M NaCl with 0.05% Sodium Azide
Mouse
Protein A purified
RUO
Primary
Protein A purified
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only