missing translation for 'onlineSavingsMsg'
Learn More

(+)-Clopidogrel hydrogen sulfate, Tocris Bioscience™

Selective P2Y12 antagonist

Brand:  Tocris (Bio-Techne) 182050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460074

  • £665.57 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective, high affinity P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity. Active enantiomer of (&plusmin;)-clopidogrel hydrochloride (Cat. No. 2490).
Specifications

Chemical Identifiers

120202-66-6
419.891
clopidogrel bisulfate, clopidogrel hydrogen sulfate, iscover, plavix, clopidogrel hemisulfate, s-methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate sulfate, clopidogrel sulfate, myogrel, clopidogrel sulphate, clopidogrel bisulphate
CHEBI:3759
COC(=O)C(C1=CC=CC=C1Cl)N2CCC3=C(C2)C=CS3.OS(=O)(=O)O
C16H18ClNO6S2
FDEODCTUSIWGLK-RSAXXLAASA-N
115366
methyl (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid

Specifications

(+)-Clopidogrel hydrogen sulfate
120202-66-6
clopidogrel bisulfate, clopidogrel hydrogen sulfate, iscover, plavix, clopidogrel hemisulfate, s-methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate sulfate, clopidogrel sulfate, myogrel, clopidogrel sulphate, clopidogrel bisulphate
COC(=O)C(C1=CC=CC=C1Cl)N2CCC3=C(C2)C=CS3.OS(=O)(=O)O
419.891
CHEBI:3759
>99%
50mg
C16H18ClNO6S2
FDEODCTUSIWGLK-RSAXXLAASA-N
methyl (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
115366
419.9
Videos