missing translation for 'onlineSavingsMsg'
Learn More

(±)-Clopidogrel hydrochloride, Tocris Bioscience™

Selective P2Y12 antagonist

Brand:  Tocris (Bio-Techne) 249050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461324

  • £531.72 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective, high affinity P2Y12 receptor antagonist (pKi = 6.9 for hP2Y12 receptor). Inhibits ADP-induced platelet aggregation; potent antithrombotic agent. Requires bioactivation, unlike PSB 0739 (Cat. No. 3983). (+)-Clopidogrel hydrogen sulfate (Cat. No. 1820) also available.
Specifications

Chemical Identifiers

130209-90-4
358.277
+/--clopidogrel hydrochloride, -clopidogrel hydrochloride, 2-chlorophenyl-6,7dihydrothieno 3,2-c pyridine-5-4h-acetic acid methyl ester sulfate, methyl 2-chlorophenyl 6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate hydrochloride 1:1, methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate hydrochloride, methyl 2-4,5,6,7-tetrahydrothieno 3,2-c pyridin-5-yl-2-2-chlorophenyl acetate hydrochloride
methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;hydrochloride
C16H17Cl2NO2S
XIHVAFJSGWDBGA-UHFFFAOYSA-N
22386150
COC(=O)C(C1=CC=CC=C1Cl)N2CCC3=C(C2)C=CS3.Cl

Specifications

(±)-Clopidogrel hydrochloride
130209-90-4
+/--clopidogrel hydrochloride, -clopidogrel hydrochloride, 2-chlorophenyl-6,7dihydrothieno 3,2-c pyridine-5-4h-acetic acid methyl ester sulfate, methyl 2-chlorophenyl 6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate hydrochloride 1:1, methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate hydrochloride, methyl 2-4,5,6,7-tetrahydrothieno 3,2-c pyridin-5-yl-2-2-chlorophenyl acetate hydrochloride
COC(=O)C(C1=CC=CC=C1Cl)N2CCC3=C(C2)C=CS3.Cl
358.277
358.28
50mg
C16H17Cl2NO2S
XIHVAFJSGWDBGA-UHFFFAOYSA-N
methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;hydrochloride
22386150
>99%
Videos