missing translation for 'onlineSavingsMsg'
Learn More

Clofarabine, Tocris Bioscience™

Deoxycytidine kinase (dCK) substrate

Brand:  Tocris (Bio-Techne) 260010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13431554

  • £198.45 / 10mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Deoxycytidine kinase (dCK) substrate. Phosphorylated to form clofarabine triphosphate, which competes with dATP for DNA polymerase-α and -ε and potently inhibits ribonucleotide reductase (IC50 = 65 nM). Induces apoptosis by directly altering mitochondrial transmembrane potential. Demonstrates growth inhibition and cytotoxic activity in a variety of leukemias and solid tumors.
Specifications

Chemical Identifiers

123318-82-1
303.678
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
CHEBI:681569
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)Cl
C10H11ClFN5O3
WDDPHFBMKLOVOX-AYQXTPAHSA-N
119182
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol

Specifications

10mg
123318-82-1
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)Cl
303.678
CHEBI:681569
>99%
Clofarabine
C10H11ClFN5O3
WDDPHFBMKLOVOX-AYQXTPAHSA-N
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
119182
303.68
Videos
Documents