Learn More
Tocris Bioscience™ Clobenpropit dihydrobromide
Highly potent H3 antagonist and H4 partial agonist
Brand:  Tocris Bioscience™ 0752/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13437963
  • £174.93 / 10mg
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
An extremely potent histamine H3 antagonist/inverse agonist (pA2=9.93). Also displays partial agonist activity at H4 receptors; induces eosinophil shape change with an EC50 of 3 nM. Also available as part of the Histamine H3 Receptor Tocriset™ .
Specifications
Specifications
N-(4-Chlorobenzyl)-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
>99%
Observed MW: 470.65
Soluble to 100 mM in water
145231-35-2
C14H17ClN4S.2HBr