Learn More
Thermo Scientific™ ClipTip™ Filtered Pipette Tips (EU IVD/CE-marked)
Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
£73.44 - £95.91
Products 7
Product Code Brand Description Volume (Metric) Color Code Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Color Code Price Quantity & Availability  
15932021
View accessory products View Documents Promotion Details
Thermo Scientific™
94420063
ClipTip 12.5 Ext, filter, sterile 12.5 μL Pink
£87.95
Pack of 960
View alternative products
Save Up to 
»
In stock, ready to ship: 668
Add to basket
13276269
View accessory products View Documents Promotion Details
Thermo Scientific™
94420213
ClipTip 20, filter, sterile 20 μL Pink
£89.23
Pack of 960
View alternative products
Save Up to 
»
In stock, ready to ship: 65
Add to basket
13246269
View accessory products View Documents Promotion Details
Thermo Scientific™
94420253
ClipTip 50, filter, sterile 50 μL Violet
£89.23
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 22-02-2022
Add to basket
13286269
View accessory products View Documents Promotion Details
Thermo Scientific™
94420313
ClipTip 200, filter, sterile 200 μL Yellow
£88.73
Pack of 960
View alternative products
Save Up to 
»
In stock, ready to ship: 2517
Add to basket
13296269
View accessory products View Documents Promotion Details
Thermo Scientific™
94420513
ClipTip 300, filter, sterile 300 μL Orange
£88.73
Pack of 960
View alternative products
Save Up to 
»
In stock, ready to ship: 626
Add to basket
15952021
View accessory products View Documents Promotion Details
Thermo Scientific™
94420613
ClipTip 300 Ext, filter, sterile 300 μL Orange
£73.44
Pack of 768
View alternative products
Save Up to 
»
In stock, ready to ship: 298
Add to basket
13226279
View accessory products View Documents Promotion Details
Thermo Scientific™
94420713
ClipTip 1000, filter, sterile 1000 μL Blue
£95.91
Pack of 768
View alternative products
Save Up to 
»
In stock, ready to ship: 45
Description
Description
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.
Key Features:
  • ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
  • Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Low retention
  • Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.
Proven quality
  • Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
Environmentally responsible
  • ClipTip reload system uses recycled components and 30% less plastic
Versatile Tip Range
  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250μL
  • 384-format volumes: 12.5, 30 and 125µL
  • Extended length tips available for volumes 12.5 and 300 µl allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Warranty
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.