Learn More
Nickel Electro™ Energy Regulator
Nickel Electro™ Energy Regulator for use with circular or rectangular boiling baths and enables energy to heating element to be varied allowing gentle to vigorous boil.
Brand:  Nickel Electro™ ER-1
Code : 03
Additional Details : Weight : 1.00000kg
Product Code. 12758908
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Adds level of boiling control to Circular and Rectangular, multiplace series of Boiling Baths.
  • Simple to use control, robust and reliable
  • Small footprint
  • Non slip feet
  • The unit is fitted with an energy regulator which controls the amount of boiling in a bath from a gentle simmer to a vigorous boil.
  • Often used to reduce the amount of energy consumed by boiling bath, which is constantly boiling and demanding power, using this accessory which regulates power demand.
Specifications
Specifications
Circular and Rectangular, Multi-place Series of Boiling Baths
128 mm
1.5 kg
50/60 Hz
Energy Regulator
125 mm
230 V