missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Click-On™ Inline Gas Filter Connectors

Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters.

Brand:  Thermo Scientific™ 60180-809

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10088952

  • £129.00 / Pack of 2


Estimated Shipment: 12-11-2021
Add to basket

Description

Description

Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters. The end fittings remain installed during filter replacement, thereby reducing the cost of clean gas.

  • Incorporates needle valves that close lines and prevent contamination
  • End fittings measure 0.125in. (0.3cm) or 0.25in. (0.6cm)
  • Reusable
Specifications

Specifications

0.125 in.
Click-On Inline Filter
Brass
Brass End Fitting
Videos
SDS
Documents
Certificates

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.