missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Click-On™ Inline Gas Filter Connectors

Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters.

Brand:  Thermo Scientific™ 60180-812

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10340925

  • £338.08 / Pack of 2
In Stock
to see stock.
Add to basket

Description

Description

Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters. The end fittings remain installed during filter replacement, thereby reducing the cost of clean gas.

  • Incorporates needle valves that close lines and prevent contamination
  • End fittings measure 0.125in. (0.3cm) or 0.25in. (0.6cm)
  • Reusable
Specifications

Specifications

Click-On Inline Filter
0.25 in.
Steel
Steel End Fitting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.