missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC3B/Tetranectin Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | CLEC3B/Tetranectin |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452 |
Gene ID (Entrez) | 7123 |
Gene Accession No. | P05452 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
CLEC3B/Tetranectin Polyclonal specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Western Blot.Specifications
CLEC3B/Tetranectin | |
7123 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human CLEC3B/Tetranectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) CLEC1, rhCLEC2, rhCLEC10A, and rhCLEC14A is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452 | |
P05452 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Sheep | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only