Learn More
Diversey™ ClearKlens Tego 2000 Disinfectant
Cleans and disinfects in sterile environments. The Diversey ClearKlens Tego 2000 Disinfectant is an active amphoteric disinfectant with a broad spectrum of activity. This microbiocidal disinfectant can be applied by means of a mop or cloth.
Brand:  Diversey™ 7516051
Code : Z3
Additional Details : CAS Number : 139734-65-9 Weight : 20.00000kg
 View more versions of this product
Product Code. 15370318
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Hard surface disinfectant based on amphoretics
  • Supplied with certificates of analysis, sterility, and irradiation where applicable
Specifications
Specifications
RTU VH25S
20L
≈8 (neat)
4 x 5L
Sterile