Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates
Carry out colormetric assays with reliable and reproducible results using these clear, flat-bottom, immuno nonsterile 384-well plates, offered with a choice of surface.
Brand: Thermo Scientific™ 8755
Code : NEW
Additional Details : Weight : 3.35700kg
Alert:
Each lot is functionally tested for IgG binding.
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, offered with a choice of surface for optimal binding. Includes Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
384-Well Solid Plates:
- Clear polystyrene
- Choice of surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo ScientificMaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
No Barcode | |
Standard | |
16 x 24 | |
Adsorption | |
Microplate | |
120 μL | |
100 μL | |
Immulon 4 HBX | |
50 | |
None |
Hydrophilic | |
Colorimetric | |
50 Cases | |
Clear | |
Polystyrene | |
384 | |
Non-sterile | |
Square | |
10 | |
Antibodies (Glycoproteins) |