missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Weights with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.

Brand:  Zwiebel™ PBI23726

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11794744

  • £785.46 / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Class M1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications

Specifications

Cast Iron
50 kg
±2500 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers