Learn More
Zwiebel™ Class M1 Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
Brand: Zwiebel™ PBI56845
Code : A0
Additional Details : Weight : 1.53900kg
Description
Specifications
1 to 500 g | |
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |