Learn More
Zwiebel™ Class F2 Box Weights
Brand: Zwiebel™ PBI56827
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
2 g | |
±0.4 mg | |
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |