Learn More
Zwiebel™ Class F2 Box Weights
Brand: Zwiebel™ PBI5682H
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
2000 g | |
±30 mg | |
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |