missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class F1 Box Weights with Certificate
Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.
Brand: Zwiebel™ PBI23141
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Class F1 calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
Specifications
10,000 g | |
±50 mg | |
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |