Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.
Brand: Zwiebel™ PBI23874
Code : A0
Additional Details : Weight : 0.80000kg
Description
Specifications
5 g | |
±0.05 mg | |
For use with Weighing Instruments of Special Accuracy Class I |