Learn More
VITLAB™ Class B SAN Short Form Measuring Cylinders
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B SAN Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
Brand: VITLAB™ 64091
Code : NEW
Additional Details : Weight : 0.01620kg
Description
- Crystal clear graduated cylinder
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded